MAT047025949 Matrix Scientific

2-(1,1-Dioxo-tetrahydro-1lambda*6*-thiophen-3-yl-amino)-4-methylsulfanylbutyric acid

  • MDL number MFCD00649172
  • Cas Number : 1396965-21-1
  • Molecular Weight : 267.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/007745.html

Safety Information

MSDS http://www.matrixscientific.com/007745.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart