MAT047025961 Matrix Scientific

2-(3-Methoxyphenylcarbamoyl)-cyclohexanecarboxylic acid

  • MDL number MFCD00683438
  • Cas Number : 301352-85-2
  • Molecular Weight : 277.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/007756.html

Safety Information

MSDS http://www.matrixscientific.com/007756.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart