MAT047025983 Matrix Scientific

4-(5-Methoxy-1,2-dimethyl-1H-indol-3-yl)thiazol-2-ylamine

  • MDL number MFCD00425556
  • Cas Number : 296264-57-8
  • Molecular Weight : 273.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/007776.html

Safety Information

MSDS http://www.matrixscientific.com/007776.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart