MAT047025985 Matrix Scientific

7-(4-Methyl-thiazol-2-ylsulfanyl)-heptanoic acid

  • MDL number MFCD00475038
  • Cas Number : 40107-51-5
  • Molecular Weight : 259.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/007778.html

Safety Information

MSDS http://www.matrixscientific.com/007778.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart