MAT047025993 Matrix Scientific

2-(4-Fluorobenzoyl)aminopropionic acid

  • MDL number MFCD00706946
  • Cas Number : 451-28-5
  • Molecular Weight : 211.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/007784.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/007784.html
Availability details for
Quantity
  • Availability
  • Add to Cart