MAT047025997 Matrix Scientific

3-(Naphthalene-2-sulfonyl)-thiazolidine-2-carboxylic acid

  • MDL number MFCD00836469
  • Cas Number : 436811-27-7
  • Molecular Weight : 323.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/007788.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/007788.html
Availability details for
Quantity
  • Availability
  • Add to Cart