MAT047026002 Matrix Scientific

1-(3-Dimethylaminopropyl)-1H-benzoimidazol-2-yl-amine

  • MDL number MFCD01055040
  • Cas Number : 38652-80-1
  • Molecular Weight : 218.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/007792.html

Safety Information

MSDS http://www.matrixscientific.com/007792.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart