MAT047026008 Matrix Scientific

3-Acetyl-2-(4-fluoro-phenyl)-thiazolidine-4-carboxylic acid

  • MDL number MFCD00613804
  • Cas Number : 312714-09-3
  • Molecular Weight : 269.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/007798.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/007798.html
Availability details for
Quantity
  • Availability
  • Add to Cart