MAT047026009 Matrix Scientific

N-(1-Phenyl-4,5-dihydro-1H-pyrazol-3-yl)-phthalamic acid

  • MDL number MFCD01131669
  • Cas Number : 304661-57-2
  • Molecular Weight : 309.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/007799.html

Safety Information

MSDS http://www.matrixscientific.com/007799.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart