MAT047026019 Matrix Scientific

3-(3,4-Dimethoxy-benzoylamino)-3-(4-fluoro-phenyl)-propionic acid

  • MDL number MFCD01366676
  • Cas Number : 332052-63-8
  • Molecular Weight : 347.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/007808.html

Safety Information

MSDS http://www.matrixscientific.com/007808.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart