MAT047026022 Matrix Scientific

5-Fluoro-2-piperidin-1-yl-pyrimidin-4-ylamine

  • MDL number MFCD01140299
  • Cas Number : 247225-85-0
  • Molecular Weight : 196.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/007810.html

Safety Information

MSDS http://www.matrixscientific.com/007810.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart