MAT047026035 Matrix Scientific

5-(4-Chloro-phenyl)-1H-pyrrole-2-carboxylicacid

  • MDL number MFCD01217348
  • Cas Number : 41019-45-8
  • Molecular Weight : 221.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 182-186
Country of Origin USA
Product Link http://www.matrixscientific.com/007822.html

Safety Information

MSDS http://www.matrixscientific.com/007822.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart