MAT047026045 Matrix Scientific

5,6-Dihydro-benzo[c]acridine-7-carboxylic acid

  • MDL number MFCD01860801
  • Cas Number : 83-93-2
  • Molecular Weight : 275.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 250
Country of Origin USA
Product Link http://www.matrixscientific.com/007831.html

Safety Information

MSDS http://www.matrixscientific.com/007831.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart