MAT047026066 Matrix Scientific

[4-Ethyl-5-(4-hydroxyphenyl)-4H-[1,2,4]triazol-3-ylsulfanyl]-acetic acid

  • MDL number MFCD02226921
  • Cas Number : 462066-81-5
  • Molecular Weight : 279.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/007850.html

Safety Information

MSDS http://www.matrixscientific.com/007850.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart