MAT047026073 Matrix Scientific

1-(4-Propoxybenzoyl)-pyrrolidine-2-carboxylicacid

  • MDL number MFCD00423324
  • Cas Number : 1246471-39-5
  • Molecular Weight : 277.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/007857.html

Safety Information

MSDS http://www.matrixscientific.com/007857.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart