MAT047026078 Matrix Scientific

2-(2-Amino-thiazol-4-yl)-N-(2-methoxy-phenyl)-acetamide

  • MDL number MFCD02742558
  • Cas Number : 333421-22-0
  • Molecular Weight : 263.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/007861.html

Safety Information

MSDS http://www.matrixscientific.com/007861.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart