MAT047026091 Matrix Scientific

Benzyl-(2-chloro-benzyl)-amine

  • MDL number MFCD00657915
  • Cas Number : 67342-76-1
  • Molecular Weight : 231.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Boiling point 194-195� Torr
Product Link http://www.matrixscientific.com/007873.html

Safety Information

MSDS http://www.matrixscientific.com/007873.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart