MAT047026104 Matrix Scientific

5-[2-(Toluene-4-sulfonyl)ethyl]-[1,3,4]-thiadiazol-2-ylamine

  • MDL number MFCD00436809
  • Cas Number : 302956-47-4
  • Molecular Weight : 283.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/007884.html

Safety Information

MSDS http://www.matrixscientific.com/007884.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart