MAT047026109 Matrix Scientific

[(6-Amino-9H-purin-8-yl)sulfanyl]acetic acid

  • MDL number MFCD00453651
  • Cas Number : 304444-52-8
  • Molecular Weight : 225.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/007889.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/007889.html
Availability details for
Quantity
  • Availability
  • Add to Cart