MAT047026122 Matrix Scientific

(1,1-Dioxo-2,3-dihydro-1H-1lambda*6*-thiophen-3-yl-amino)acetic acid

  • MDL number MFCD00195978
  • Cas Number : 201990-24-1
  • Molecular Weight : 191.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/007900.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/007900.html
Availability details for
Quantity
  • Availability
  • Add to Cart