MAT047026138 Matrix Scientific

1-(2,4-Dimethylphenoxy)-3-methylamino-propan-2-ol

  • MDL number MFCD00449241
  • Cas Number : 414886-67-2
  • Molecular Weight : 209.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/007915.html

Safety Information

MSDS http://www.matrixscientific.com/007915.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart