MAT047026157 Matrix Scientific

3-(4-Fluorophenylcarbamoyl)-1,2,2-trimethyl-cyclopentanecarboxylic acid

  • MDL number MFCD00554930
  • Cas Number : 304666-33-9
  • Molecular Weight : 293.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 180-181
Country of Origin USA
Product Link http://www.matrixscientific.com/007932.html

Safety Information

MSDS http://www.matrixscientific.com/007932.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart