MAT047026186 Matrix Scientific

(Pyridin-2-ylsulfanyl)acetic acid

  • MDL number MFCD00276114
  • Cas Number : 10002-29-6
  • Molecular Weight : 169.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 127-133
Country of Origin USA
Product Link http://www.matrixscientific.com/007960.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/007960.html
Availability details for
Quantity
  • Availability
  • Add to Cart