MAT047026187 Matrix Scientific

3-Amino-5-fluorobenzotrifluoride

  • MDL number MFCD00061315
  • Cas Number : 454-67-1
  • Molecular Weight : 179.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/001698.html

Safety Information

hcodes Irritant/Refrigerate
MSDS http://www.matrixscientific.com/001698.html
Availability details for
Quantity
  • Availability
  • Add to Cart