MAT047026191 Matrix Scientific

N-(2,6-Dimethoxypyrimidin-4-yl)succinamic acid

  • MDL number MFCD00978708
  • Cas Number : 303134-31-8
  • Molecular Weight : 255.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/007964.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/007964.html
Availability details for
Quantity
  • Availability
  • Add to Cart