MAT047026199 Matrix Scientific

3-(5-Oxo-2-thioxoimidazolidin-4-yl)-propionic acid

  • MDL number MFCD01151881
  • Cas Number : 83178-70-5
  • Molecular Weight : 188.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 122
Country of Origin USA
Product Link http://www.matrixscientific.com/007971.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/007971.html
Availability details for
Quantity
  • Availability
  • Add to Cart