MAT047026202 Matrix Scientific

2-(3,5-Dioxo-4-aza-tricyclo[5.2.1.0*2,6*]-dec-4-yl)propionic acid

  • MDL number MFCD06799406
  • Cas Number : 32392-58-8
  • Molecular Weight : 237.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/007974.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/007974.html
Availability details for
Quantity
  • Availability
  • Add to Cart