MAT047026290 Matrix Scientific

2-(Trifluoromethoxy)chlorobenzene

  • MDL number MFCD03840206
  • Cas Number : 450-96-4
  • Molecular Weight : 196.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Density 1.369/20
Country of Origin USA
Boiling point 142-143
Product Link http://www.matrixscientific.com/008763.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/008763.html
Availability details for
Quantity
  • Availability
  • Add to Cart