MAT047026313 Matrix Scientific

3,3-Dichloro-1,1,1-trifluoroacetone trihydrate

  • MDL number MFCD04038282
  • Cas Number : 126266-75-9
  • Molecular Weight : 234.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Country of Origin USA
Boiling point 75-76
Product Link http://www.matrixscientific.com/008909.html

Safety Information

hcodes Irritant/Irritant-Harmful
MSDS http://www.matrixscientific.com/008909.html
Availability details for
Quantity
  • Availability
  • Add to Cart