MAT047026326 Matrix Scientific

[2-(1H-Indol-3-yl)-ethyl]-(3-methoxy-benzyl)-amine

  • MDL number MFCD01468856
  • Cas Number : 435342-06-6
  • Molecular Weight : 280.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/008047.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/008047.html
Availability details for
Quantity
  • Availability
  • Add to Cart