MAT047026338 Matrix Scientific

2-Methoxy-5-phenylmethanesulfonyl-phenylamine

  • MDL number MFCD00725362
  • Cas Number : 2815-50-1
  • Molecular Weight : 277.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/008058.html

Safety Information

MSDS http://www.matrixscientific.com/008058.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart