MAT047026361 Matrix Scientific

8-Methyl-10-oxo-2,3,5,6-tetrahydro-1H,4H,10H-11-oxa-3a-aza-benzo[de]anthracene-9-carbaldehyde

  • MDL number MFCD01076737
  • Cas Number : 142730-53-8
  • Molecular Weight : 283.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/008079.html

Safety Information

MSDS http://www.matrixscientific.com/008079.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart