MAT047026380 Matrix Scientific

3-(1-Methyl-1H-benzoimidazol-2-ylsulfanyl)-propionic acid

  • MDL number MFCD01788343
  • Cas Number : 342013-63-2
  • Molecular Weight : 236.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/008095.html

Safety Information

MSDS http://www.matrixscientific.com/008095.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart