MAT047026399 Matrix Scientific

3-Imidazo[1,2-a]pyridin-2-yl-phenylamine

  • MDL number MFCD00702245
  • Cas Number : 313231-71-9
  • Molecular Weight : 209.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/008112.html

Safety Information

MSDS http://www.matrixscientific.com/008112.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart