MAT047026405 Matrix Scientific

3-Hydroxy-2-(toluene-4-sulfonylamino)-butyric acid

  • MDL number MFCD00447114
  • Cas Number : 91280-33-0
  • Molecular Weight : 273.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/008118.html

Safety Information

MSDS http://www.matrixscientific.com/008118.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart