MAT047026423 Matrix Scientific

(4-Fluoro-benzyl)-(4-methoxy-benzyl)-amine

  • MDL number MFCD01134878
  • Cas Number : 355815-47-3
  • Molecular Weight : 245.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/008134.html

Safety Information

MSDS http://www.matrixscientific.com/008134.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart