MAT047026434 Matrix Scientific

2-(4-Methoxybenzoyl)benzenecarboxylic acid

  • MDL number MFCD00278217
  • Cas Number : 1151-15-1
  • Molecular Weight : 256.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 146
Country of Origin USA
Product Link http://www.matrixscientific.com/008144.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/008144.html
Availability details for
Quantity
  • Availability
  • Add to Cart