MAT047026445 Matrix Scientific

3-Chloro-4-(4-methyl-piperazin-1-yl)-phenylamine

  • MDL number MFCD01951505
  • Cas Number : 16154-72-6
  • Molecular Weight : 225.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 141
Country of Origin USA
Product Link http://www.matrixscientific.com/008154.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/008154.html
Availability details for
Quantity
  • Availability
  • Add to Cart