MAT047026470 Matrix Scientific

4-[N'-(4,6-Dimethyl-pyrimidin-2-yl)-guanidino]-benzoic acid

  • MDL number MFCD01661290
  • Cas Number : 354994-01-7
  • Molecular Weight : 285.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 253
Country of Origin USA
Product Link http://www.matrixscientific.com/008177.html

Safety Information

MSDS http://www.matrixscientific.com/008177.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart