MAT047026498 Matrix Scientific

2-Methyl-3-(3-methyl-pyrazol-1-yl)-propionic acid

  • MDL number MFCD02732635
  • Cas Number : 371212-60-1
  • Molecular Weight : 168.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 103-104
Country of Origin USA
Product Link http://www.matrixscientific.com/008201.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/008201.html
Availability details for
Quantity
  • Availability
  • Add to Cart