MAT047026499 Matrix Scientific

4-(1-Amino-3-methylsulfanyl-propyl)-hepta-1,6-dien-4-ol

  • MDL number MFCD02105248
  • Cas Number : 315249-26-4
  • Molecular Weight : 215.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/008202.html

Safety Information

MSDS http://www.matrixscientific.com/008202.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart