MAT047026515 Matrix Scientific

5-Methyl-4-(2-thienyl)-1,3-thiazol-2-amine

  • MDL number MFCD02734128
  • Cas Number : 206555-52-4
  • Molecular Weight : 196.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/008217.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/008217.html
Availability details for
Quantity
  • Availability
  • Add to Cart