MAT047026523 Matrix Scientific

N-(2-Methoxy-benzyl)-N',N'-dimethyl-benzene-1,4-diamine

  • MDL number MFCD03044444
  • Cas Number : 436088-37-8
  • Molecular Weight : 256.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/008224.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/008224.html
Availability details for
Quantity
  • Availability
  • Add to Cart