MAT047026543 Matrix Scientific

3-Cyclohexylaminomethyl-7-methoxy-1H-quinolin-2-one

  • MDL number MFCD02592004
  • Cas Number : 462068-04-8
  • Molecular Weight : 286.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/008242.html

Safety Information

MSDS http://www.matrixscientific.com/008242.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart