MAT047026567 Matrix Scientific

3,5-Dimethyl-1H-pyrazol-4-ylamine

  • MDL number MFCD00187666
  • Cas Number : 5272-86-6
  • Molecular Weight : 111.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 204-205
Country of Origin USA
Product Link http://www.matrixscientific.com/008264.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/008264.html
Availability details for
Quantity
  • Availability
  • Add to Cart