MAT047026610 Matrix Scientific

4-(2,4-Dimethyl-phenylamino)-butan-1-ol

  • MDL number MFCD02603507
  • Cas Number : 356086-89-0
  • Molecular Weight : 193.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/008302.html

Safety Information

MSDS http://www.matrixscientific.com/008302.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart