MAT047026611 Matrix Scientific

4-(4-Ethoxy-phenylamino)-butan-1-ol

  • MDL number MFCD03110163
  • Cas Number : 356539-16-7
  • Molecular Weight : 209.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/008303.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/008303.html
Availability details for
Quantity
  • Availability
  • Add to Cart