MAT047026613 Matrix Scientific

4-Benzylamino-butan-1-ol

  • MDL number MFCD00191609
  • Cas Number : 59578-63-1
  • Molecular Weight : 179.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Boiling point 120`-125�6 Torr
Product Link http://www.matrixscientific.com/008305.html

Safety Information

MSDS http://www.matrixscientific.com/008305.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart