MAT047026614 Matrix Scientific

4-(3-Methoxy-phenylamino)-butan-1-ol

  • MDL number MFCD03119380
  • Cas Number : 356539-28-1
  • Molecular Weight : 195.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/008306.html

Safety Information

MSDS http://www.matrixscientific.com/008306.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart