MAT047026633 Matrix Scientific

2-Benzoyl-1,2,3,4-tetrahydro-isoquinoline-3-carboxylic acid

  • MDL number MFCD00532902
  • Cas Number : 93316-40-6
  • Molecular Weight : 281.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 160-163
Country of Origin USA
Product Link http://www.matrixscientific.com/008323.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/008323.html
Availability details for
Quantity
  • Availability
  • Add to Cart